siRNA Details
| Gene Symbol: GAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007970.1 | XP_016863459.1 | 1 | 1299 | ||
| XM_017007971.1 | XP_016863460.1 | 1 | 1299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAB1| Gene Symbol: GAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007970.1 | XP_016863459.1 | 1 | 1299 | ||
| XM_017007971.1 | XP_016863460.1 | 1 | 1299 | |||