siRNA Details
| Gene Symbol: LOC101927766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109891.1 | 1 | 981 | |||
| GenBank mRNA | AL833484.1 | - | 1 | 981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927766| Gene Symbol: LOC101927766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109891.1 | 1 | 981 | |||
| GenBank mRNA | AL833484.1 | - | 1 | 981 | ||