siRNA Details
| Gene Symbol: ZNF518A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017016986.1 | XP_016872475.1 | 1 | 171 | ||
| XM_017016987.1 | XP_016872476.1 | 1 | 171 | |||
| XM_017016988.1 | XP_016872477.1 | 1 | 171 | |||
| XM_017016990.1 | XP_016872479.1 | 1 | 171 | |||
| XM_017016991.1 | XP_016872480.1 | 1 | 171 | |||
| XM_017016992.1 | XP_016872481.1 | 1 | 171 | |||
| XM_017016995.1 | XP_016872484.1 | 1 | 171 | |||
| XM_017016996.1 | XP_016872485.1 | 1 | 171 | |||
| GenBank mRNA | AK095317.1 | - | 1 | 244 | ||

ZNF518A
Genomic Map