siRNA Details
| Gene Symbol: FLJ31945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056507.1 | - | 1 | 565 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF279660.3 | - | 6 | 129325 | Chr. 13 - 49996975 - 50344526 [+] on Build GRCh38 | |

FLJ31945
Genomic Map