siRNA Details
| Gene Symbol: LOC100505530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126057.1 | 4 | 1172 | |||
| GenBank mRNA | AK127725.1 | - | 4 | 1172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505530| Gene Symbol: LOC100505530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126057.1 | 4 | 1172 | |||
| GenBank mRNA | AK127725.1 | - | 4 | 1172 | ||