siRNA Details
| Gene Symbol: LOC100505530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126057.1 | 2 | 383 | |||
| GenBank mRNA | AK127725.1 | - | 2 | 383 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA756763.1 | - | 2 | 383 | Chr. 6 - 37507348 - 37535616 [+] on Build GRCh38 | |

LOC100505530
Genomic Map