siRNA Details
| Gene Symbol: LINC01314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120317.1 | 3 | 6618 | |||
| GenBank mRNA | AL109671.1 | - | 1 | 2003 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DT220719.1 | - | 1 | 540 | Chr. 15 - 80195715 - 80196343 [-] on Build GRCh38 | |

LINC01314
Genomic Map