siRNA Details
| Gene Symbol: MLXIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014938.5 | NP_055753.3 | 17 | 7132 | ||
| XM_006719292.3 | XP_006719355.1 | 17 | 7119 | |||
| GenBank mRNA | BM007462.1 | - | 1 | 416 | ||
| BM857835.1 | - | 1 | 42 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLXIP| Gene Symbol: MLXIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014938.5 | NP_055753.3 | 17 | 7132 | ||
| XM_006719292.3 | XP_006719355.1 | 17 | 7119 | |||
| GenBank mRNA | BM007462.1 | - | 1 | 416 | ||
| BM857835.1 | - | 1 | 42 | |||