siRNA Details
| Gene Symbol: USP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648836.1 | - | 2 | 1660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP53| Gene Symbol: USP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648836.1 | - | 2 | 1660 | ||