siRNA Details
| Gene Symbol: TTC39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC070139.1 | - | 4 | 656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC39B| Gene Symbol: TTC39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC070139.1 | - | 4 | 656 | ||