siRNA Details
| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282736.1 | NP_001269665.1 | 13 | 4459 | ||
| NM_032812.8 | NP_116201.7 | 14 | 4606 | |||
| GenBank mRNA | AK127644.1 | - | 1 | 1184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLXDC2| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282736.1 | NP_001269665.1 | 13 | 4459 | ||
| NM_032812.8 | NP_116201.7 | 14 | 4606 | |||
| GenBank mRNA | AK127644.1 | - | 1 | 1184 | ||