siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033376.1 | 1 | 101 | |||
| GenBank mRNA | AY927443.1 | - | LIT3472 AY927443 |
1 | 102 | |
| AY927444.1 | - | LIT3473 AY927444 |
1 | 99 | ||
| AY927446.1 | - | LIT3474 AY927446 |
1 | 102 | ||

LINC00222
Genomic Map