siRNA Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 10 | 3807 | ||
| NM_024735.4 | NP_079011.3 | 9 | 4059 | |||
| GenBank mRNA | BI914636.1 | - | 1 | 496 | ||
| DA493744.1 | - | 1 | 70 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR621785.1 | - | 1 | 491 | Chr. 16 - 87327016 - 87329388 [-] on Build GRCh38 | |

FBXO31
Genomic Map