siRNA Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 10 | 5527 | ||
| NM_024735.4 | NP_079011.3 | 9 | 5779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO31| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 10 | 5527 | ||
| NM_024735.4 | NP_079011.3 | 9 | 5779 | |||