siRNA Details
| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518646.2 | XP_011516948.1 | 18 | 4181 | ||
| GenBank mRNA | AK094719.1 | - | 1 | 1703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERCC6L2| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518646.2 | XP_011516948.1 | 18 | 4181 | ||
| GenBank mRNA | AK094719.1 | - | 1 | 1703 | ||