siRNA Details
| Gene Symbol: SUGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 9642 | ||
| XM_011534893.2 | XP_011533195.1 | 13 | 9360 | |||
| XM_017020365.1 | XP_016875854.1 | 14 | 9532 | |||
| XM_017020366.1 | XP_016875855.1 | 14 | 9407 | |||
| XM_017020367.1 | XP_016875856.1 | 14 | 9441 | |||
| XM_017020368.1 | XP_016875857.1 | 14 | 9453 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL831999.1 | - | 2 | 1472 | Chr. 13 - 52689587 - 52700758 [+] on Build GRCh38 | |

SUGT1
Genomic Map