siRNA Details
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 9215 | ||
XM_011534893.2 | XP_011533195.1 | 13 | 8933 | |||
XM_017020365.1 | XP_016875854.1 | 14 | 9105 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 8980 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 9014 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 9026 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL831999.1 | - | 2 | 1045 | Chr. 13 - 52689587 - 52700758 [+] on Build GRCh38 |
