siRNA Details
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 13213 | ||
XM_017020365.1 | XP_016875854.1 | 14 | 13103 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 12978 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 13012 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 13024 |
