siRNA Details
| Gene Symbol: SUGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 13161 | ||
| XM_017020365.1 | XP_016875854.1 | 14 | 13051 | |||
| XM_017020366.1 | XP_016875855.1 | 14 | 12926 | |||
| XM_017020367.1 | XP_016875856.1 | 14 | 12960 | |||
| XM_017020368.1 | XP_016875857.1 | 14 | 12972 | |||

SUGT1
Genomic Map