siRNA Details
| Gene Symbol: FAM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103537.1 | 1 | 38 | |||
| GenBank mRNA | BC037297.2 | - | 1 | 38 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG717045.1 | - | 1 | 43 | Chr. 8 - 380055 - 383174 [-] on Build GRCh38 | |

FAM87A
Genomic Map