siRNA Details
Gene Symbol: FAM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103537.1 | 3 | 1702 | |||
GenBank mRNA | BC037297.2 | - | 3 | 1702 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000330148.6 | - | 3 | 1702 | Chr. 8 - 375934 - 383174 [-] on Build GRCh38 |
