siRNA Details
| Gene Symbol: LINC01231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121585.1 | 4 | 744 | |||
| GenBank mRNA | BC041456.1 | - | 4 | 744 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI756497.1 | - | 4 | 744 | Chr. 9 - 3181589 - 3198525 [+] on Build GRCh38 | |

LINC01231
Genomic Map