siRNA Details
| Gene Symbol: LOC101928988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120344.1 | 1 | 92 | |||
| GenBank mRNA | BC039388.1 | - | 1 | 92 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI562679.1 | - | 1 | 97 | Chr. 15 - 63928274 - 63935878 [+] on Build GRCh38 | |

LOC101928988
Genomic Map