siRNA Details
| Gene Symbol: CTD-2194D22.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109912.1 | 4 | 1536 | |||
| GenBank mRNA | BC039383.1 | - | 4 | 1535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-2194D22.4| Gene Symbol: CTD-2194D22.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109912.1 | 4 | 1536 | |||
| GenBank mRNA | BC039383.1 | - | 4 | 1535 | ||