siRNA Details
Gene Symbol: CTD-2194D22.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109912.1 | 4 | 938 | |||
GenBank mRNA | BC039383.1 | - | 4 | 937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CTD-2194D22.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109912.1 | 4 | 938 | |||
GenBank mRNA | BC039383.1 | - | 4 | 937 |