siRNA Details
| Gene Symbol: CTD-2194D22.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109912.1 | 3 | 133 | |||
| GenBank mRNA | BC039383.1 | - | 3 | 132 | ||
| BI463559.1 | - | 3 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-2194D22.4| Gene Symbol: CTD-2194D22.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109912.1 | 3 | 133 | |||
| GenBank mRNA | BC039383.1 | - | 3 | 132 | ||
| BI463559.1 | - | 3 | 137 | |||