siRNA Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 4770 | ||
| XM_005271139.3 | XP_005271196.1 | 5 | 4718 | |||
| GenBank mRNA | AK094134.1 | - | 1 | 2056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFP69B| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 4770 | ||
| XM_005271139.3 | XP_005271196.1 | 5 | 4718 | |||
| GenBank mRNA | AK094134.1 | - | 1 | 2056 | ||