siRNA Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 3850 | ||
| XM_005271139.3 | XP_005271196.1 | 5 | 3798 | |||
| GenBank mRNA | AK094134.1 | - | 1 | 1136 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA801448.1 | - | 1 | 80 | Chr. 1 - 40465375 - 40465946 [+] on Build GRCh38 | |

ZFP69B
Genomic Map