siRNA Details
| Gene Symbol: BTD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006713314.3 | XP_006713377.1 | 5 | 6891 | ||
| XM_011534041.2 | XP_011532343.1 | 5 | 6728 | |||
| XM_017007088.1 | XP_016862577.1 | 6 | 7004 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK096254.1 | - | 1 | 2802 | Chr. 3 - 15647560 - 15650437 [+] on Build GRCh38 | |

BTD
Genomic Map