siRNA Details
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515150.2 | XP_011513452.1 | 7 | 7982 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127523.1 | - | 1 | 1217 | Chr. 7 - 2717833 - 2727060 [+] on Build GRCh38 |
