siRNA Details
| Gene Symbol: USP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011511398.2 | XP_011509700.1 | 28 | 3327 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR611364.1 | - | 1 | 949 | Chr. 2 - 233487681 - 233488898 [-] on Build GRCh38 | |

USP40
Genomic Map