siRNA Details
| Gene Symbol: FENDRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036444.1 | 4 | 2526 | |||
| GenBank mRNA | AK097312.1 | - | 1 | 2437 | ||
| AL833740.1 | - | 1 | 2109 | |||
| BX538111.1 | - | 2 | 2726 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM922208.1 | - | 1 | 835 | Chr. 16 - 86487715 - 86488631 [-] on Build GRCh38 | |

FENDRR
Genomic Map