siRNA Details
| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120478.1 | 1 | 197 | |||
| GenBank mRNA | AY387665.1 | - | 1 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129940| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120478.1 | 1 | 197 | |||
| GenBank mRNA | AY387665.1 | - | 1 | 197 | ||