siRNA Details
| Gene Symbol: LOC101928797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109806.1 | 6 | 967 | |||
| GenBank mRNA | BC034724.1 | - | 6 | 967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928797| Gene Symbol: LOC101928797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109806.1 | 6 | 967 | |||
| GenBank mRNA | BC034724.1 | - | 6 | 967 | ||