siRNA Details
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006722488.2 | XP_006722551.1 | 18 | 3539 | ||
| XM_011526036.1 | XP_011524338.1 | 18 | 3562 | |||
| XM_011526037.1 | XP_011524339.1 | 18 | 3564 | |||
| XM_011526038.1 | XP_011524340.1 | 18 | 3559 | |||
| XM_011526040.1 | XP_011524342.1 | 19 | 3563 | |||
| XM_011526041.1 | XP_011524343.1 | 17 | 3379 | |||
| XM_017025795.1 | XP_016881284.1 | 18 | 3561 | |||
| XM_017025796.1 | XP_016881285.1 | 19 | 3573 | |||
| XM_017025800.1 | XP_016881289.1 | 16 | 3258 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR610892.1 | - | 1 | 71 | Chr. 18 - 49041501 - 49043051 [-] on Build GRCh38 | |

DYM
Genomic Map