siRNA Details
| Gene Symbol: LINC00424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047040.1 | 2 | 181 | |||
| GenBank mRNA | AK057351.1 | - | 3 | 1325 | ||
| BX092749.1 | - | 2 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00424| Gene Symbol: LINC00424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047040.1 | 2 | 181 | |||
| GenBank mRNA | AK057351.1 | - | 3 | 1325 | ||
| BX092749.1 | - | 2 | 181 | |||