siRNA Details
| Gene Symbol: DCTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199743.1 | NP_001186672.1 | 6 | 4419 | ||
| NM_032486.3 | NP_115875.1 | 6 | 4389 | |||
| NR_037573.1 | 7 | 4490 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022200.1 | - | 1 | 797 | Chr. 16 - 23670011 - 23673127 [+] on Build GRCh38 | |

DCTN5
Genomic Map