siRNA Details
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 6 | 4864 | ||
NM_032486.3 | NP_115875.1 | 6 | 4834 | |||
NR_037573.1 | 7 | 4935 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK022200.1 | - | 1 | 1242 | Chr. 16 - 23670011 - 23673127 [+] on Build GRCh38 |
