siRNA Details
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 6 | 5746 | ||
NM_032486.3 | NP_115875.1 | 6 | 5716 | |||
NR_037573.1 | 7 | 5817 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 6 | 5746 | ||
NM_032486.3 | NP_115875.1 | 6 | 5716 | |||
NR_037573.1 | 7 | 5817 |