siRNA Details
| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 6953 | ||
| XM_011515596.2 | XP_011513898.1 | 13 | 6469 | |||
| XM_017012746.1 | XP_016868235.1 | 14 | 6960 | |||
| XM_017012747.1 | XP_016868236.1 | 14 | 6993 | |||
| GenBank mRNA | AB073667.1 | - | 1 | 1150 | ||
| AL832700.1 | - | 1 | 2175 | |||
| CR627312.1 | - | 1 | 4693 | |||

PLEKHA8
Genomic Map