siRNA Details
| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 4273 | ||
| XM_011515596.2 | XP_011513898.1 | 13 | 3789 | |||
| XM_017012746.1 | XP_016868235.1 | 14 | 4280 | |||
| XM_017012747.1 | XP_016868236.1 | 14 | 4313 | |||
| GenBank mRNA | CR627312.1 | - | 1 | 2013 | ||

PLEKHA8
Genomic Map