siRNA Details
| Gene Symbol: SORBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008761.1 | XP_016864250.1 | 15 | 8291 | ||
| GenBank mRNA | BC030082.1 | - | 1 | 47 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SORBS2| Gene Symbol: SORBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008761.1 | XP_016864250.1 | 15 | 8291 | ||
| GenBank mRNA | BC030082.1 | - | 1 | 47 | ||