siRNA Details
| Gene Symbol: ANKRD20A19P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073430.1 | 3 | 3695 | |||
| GenBank mRNA | AK127292.1 | - | 3 | 3695 | ||
| AK128225.1 | - | 3 | 1162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD20A19P| Gene Symbol: ANKRD20A19P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073430.1 | 3 | 3695 | |||
| GenBank mRNA | AK127292.1 | - | 3 | 3695 | ||
| AK128225.1 | - | 3 | 1162 | |||