siRNA Details
| Gene Symbol: L3MBTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525762.1 | XP_011524064.1 | 17 | 3534 | ||
| GenBank mRNA | AK098363.1 | - | 1 | 1049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3MBTL4| Gene Symbol: L3MBTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525762.1 | XP_011524064.1 | 17 | 3534 | ||
| GenBank mRNA | AK098363.1 | - | 1 | 1049 | ||