siRNA Details
| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005253351.3 | XP_005253408.1 | 4 | 25838 | ||
| XM_011520628.2 | XP_011518930.1 | 14 | 28904 | |||
| XM_011520629.2 | XP_011518931.1 | 15 | 28711 | |||
| XM_017019219.1 | XP_016874708.1 | 14 | 28459 | |||
| GenBank mRNA | BX648060.1 | - | 1 | 1029 | ||

GRIN2B
Genomic Map