siRNA Details
| Gene Symbol: DLGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030096.1 | - | 1 | 468 | ||
| BC045785.1 | - | 1 | 463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLGAP1| Gene Symbol: DLGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030096.1 | - | 1 | 468 | ||
| BC045785.1 | - | 1 | 463 | |||