siRNA Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530187.2 | XP_011528489.1 | 1 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF289562.1 | - | 1 | 1993 | Chr. 22 - 39475441 - 39477566 [+] on Build GRCh38 | |

MGAT3
Genomic Map