siRNA Details
| Gene Symbol: LOC101927769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110117.1 | 5 | 815 | |||
| GenBank mRNA | AK026386.1 | - | 2 | 685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927769| Gene Symbol: LOC101927769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110117.1 | 5 | 815 | |||
| GenBank mRNA | AK026386.1 | - | 2 | 685 | ||