siRNA Details
| Gene Symbol: SPAG5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040012.1 | 2 | 167 | |||
| GenBank mRNA | BC001556.1 | - | 1 | 961 | ||
| BG723958.1 | - | 2 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPAG5-AS1| Gene Symbol: SPAG5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040012.1 | 2 | 167 | |||
| GenBank mRNA | BC001556.1 | - | 1 | 961 | ||
| BG723958.1 | - | 2 | 172 | |||