siRNA Details
| Gene Symbol: LINC00165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC009635.1 | - | 1 | 391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00165| Gene Symbol: LINC00165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC009635.1 | - | 1 | 391 | ||